Cleanroom overalls, BioClean-D™ CleanTough™, disposable
Supplier: Nitritex
Certificates
About this item
These disposable overalls made from CleanTough™ fabric provide good protection against hazardous dusts (including nuclear particles) and limited protection against chemical splashes. Available with hood or with collar.
- Lightweight, low linting and hard-wearing material
- Available as sterile or non sterile version
- Zip front with flap cover, thumb loops
- Elasticated back, cuffs and ankles
- Each size carries a unique barcode on the outer carton (ITF-14), this allows automatic data entry into stock control systems
Suitable for use in Class 10, ISO 4, M2.5 environments.
Colour: White.
Type 5/6; Cat. III
Certifications: EN ISO 13982-1:2004+A1:2009 (Type 5), EN ISO 13034:2005+A1:2010 (Type 6), EN 14325, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2; Cat. III
Manufactured in a facility holding ISO 9001:2015 and (EU) 2016/425 Module D certifications.
Sterile models: Sterilised in accordance with ISO 11137-1:2006.
Manufactured in a facility holding ISO 9001:2015 and (EU) 2016/425 Module D certifications.
Sterile models: Sterilised in accordance with ISO 11137-1:2006.
Packaging: One overall per sealed plastic bag; 20 overalls per sealed carton liner; one carton liner per carton. Sizes 3XL and 4XL are available on request.
Specifications
- Catalog No:
- 113-7996
- 113-7995
- NITRS-BDCHT-XXXXL
- 113-8196
- 113-7998
- 113-7997
- 113-8005
- 113-8018
- 113-8000
- 113-8006
- 113-8007
- 113-8013
- 113-8033
- 113-8004
- 113-8032
- 113-8008
- 114-9442
- 114-9441
- 113-8009
- 113-8011
- 113-8010
- 113-8012
- 113-8365
- 113-8017
- 113-7994
- 113-8019
- 113-8016
- Classe di cleanroom:
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- Applicazioni per abiti:
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
- Chemical protection, Cleanroom
Specifications
Related Information
Frequently Bought Together