Orders LinkContactUsLinkComponent
Wipe rolls, WYPALL* L10 EXTRA+
Wipe rolls, WYPALL* L10 EXTRA+
Wipe rolls, WYPALL* L10 EXTRA+
Wipe rolls, WYPALL* L10 EXTRA+
Wipe rolls, WYPALL* L10 EXTRA+
These single use wipes made from single ply AIRFLEX* material are ideal for multipurpose cleaning and medium duty wiping tasks such as polishing glass, smooth surfaces and cleaning tools.
  • Stronger than conventional single ply wipes
  • Strong and absorbent
  • Clean up fast without disintegrating
  • Can be used with a dispensing system to help control use and reduce wastage
Large rolls: For use with dispensers 6146, 6154 and 6155.
Centrefeed rolls: For use with dispensers 7017 and 7087.
Small rolls: For use with dispenser 7041.
Roll Control wipes: For use with dispensers 7018 and 7181.
Product Family Options
  • Descrizione
    Colore
    L×L
    Confezione
  • Rotolo grande, 1 velo
    bianco
    385×325 mm
    1 rotolo da 1000 panni
    1 * 1 ROU
    1 * 1 ROU - SFr. 83.80
    SFr. 83.80
    1 * 1 ROU
       SFr. 83.80
  • Rotolo grande, velo singolo
    bianco
    385×235 mm
    1 rotolo da 1000 panni
    1 * 1 ROU
    1 * 1 ROU - SFr. 81.80
    SFr. 81.80
    1 * 1 ROU
       SFr. 81.80
  • Rotolo grande, velo singolo
    blu
    385×325 mm
    1 rotolo da 1000 panni
    1 * 1 ROU
    1 * 1 ROU - SFr. 83.80
    SFr. 83.80
    1 * 1 ROU
       SFr. 83.80
  • Rotolo piccolo, velo singolo
    blu
    460×240 mm
    24 rotoli da 115 strappi ciascuno
    1 * 24 ROU
    1 * 24 ROU
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.