TPE buffer, 10X tris-phosphate-EDTA, pH 8
Catalog # ROCKMB-021
Supplier: Rockland Immunochemicals
CAS Number:
Specifications
- Conf:1 L
- Descrizione:Tris-phosphate-EDTA 10X (TPE) (0,9 M tris-phosphate 0,01 M EDTA pH 8,0)
- Seq:10
Specifications
About this item
This product is a concentrated stock solution and should be diluted appropriately with distilled, deionised water or equivalent to its final working concentration. 10X Tris-Phosphate EDTA (TPE) consists of 0,9 M Tris-Phosphate, 0,02 M EDTA at a pH of 8,0. Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionsed water.
This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.