- Tipo anticorpo:Primary
- Tipo di anticorpi:CCR4-NOT transcription complex, subunit 8
- Simbolo dell'antigene:CNOT8
- Clonalità:Policlonale
- Coniugazione:FITC (Fluoresceina Isotiocionato)
- Ospite:Rabbit
- ImmunoChimica:Yes
- Isotipo:IgG
- Reattività:Human,Rat,Mouse
- Assorbimento incrociato:No
- Forma:liquid
- ID gene:9337
- Sinomini antigene:hCAF1|CALIF|Caf1b|POP2|CAF1
- Tampone di conservazione:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Temperatura di conservazione:Store at –20 °C for 12 months
- Concentrazione:1 μg/μl
- Temperatura di spedizione:4°C
- Purificazione:Purified by Protein A
- Conf:100 µl
The CCR4-NOT complex is an evolutionarily conserved, multi-component complex known to be involved in transcription as well as mRNA degradation. Various subunits within the complex are involved in influencing nuclear hormone receptor activities. The CCR4-NOT complex is also involved in the regulation of Histone H3 lysine 4 methylation through a ubiquitin-dependent pathway that likely involves the proteasome. CNOT8 (CCR4-NOT transcription complex subunit 8), also known as CALIF or POP2, is a 292 amino acid protein that localizes to both the nucleus and the cytoplasm and functions as part of the CCR-NOT complex. Expressed ubiquitously, CNOT8 plays a role in transcriptional regulation for a diverse set of processes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CNOT8
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: