Order Entry
Switzerland
ContactUsLinkComponent
Anti-RAB5 Rabbit Polyclonal Antibody (Cy3®)
Anti-RAB5 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-6175R-CY3
Supplier:  Bioss
Anti-RAB5 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-6175R-CY3
Supplier:  Bioss

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Ras-related protein Rab-5
  • Simbolo dell'antigene:
    RAB5
  • Clonalità:
    Policlonale
  • Coniugazione:
    Cy3®
  • Ospite:
    Rabbit
  • ImmunoChimica:
    Yes
  • Isotipo:
    IgG
  • Reattività:
    Human,
    Rat,
    Mouse
  • Assorbimento incrociato:
    No
  • Forma:
    liquid
  • ID gene:
    5868
  • Sinomini antigene:
    RAB5A|Rab5a|RAB 5|RAS|RAB 5A|RAB5|RAS associated protein
  • Tampone di conservazione:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Temperatura di conservazione:
    Store at 4°C for 12 months
  • Concentrazione:
    1 μg/μl
  • Temperatura di spedizione:
    4°C
  • Purificazione:
    Purified by Protein A
  • Conf:
    100 µl

Specifications

About this item

Rab5-related subfamily. This subfamily includes Rab5 and Rab22 of mammals, Ypt51/Ypt52/Ypt53 of yeast, and RabF of plants. The members of this subfamily are involved in endocytosis and endocytic-sorting pathways. In mammals, Rab5 GTPases localize to early endosomes and regulate fusion of clathrin-coated vesicles to early endosomes and fusion between early endosomes. In yeast, Ypt51p family members similarly regulate membrane trafficking through prevacuolar compartments. GTPase activating proteins (GAPs) interact with GTP-bound Rab and accelerate the hydrolysis of GTP to GDP. Guanine nucleotide exchange factors (GEFs) interact with GDP-bound Rabs to promote the formation of the GTP-bound state. Rabs are further regulated by guanine nucleotide dissociation inhibitors (GDIs), which facilitate Rab recycling by masking C-terminal lipid binding and promoting cytosolic localization. Most Rab GTPases contain a lipid modification site at the C-terminus, with sequence motifs CC, CXC, or CCX. Lipid binding is essential for membrane attachment, a key feature of most Rab proteins. Due to the presence of truncated sequences in this CD, the lipid modification site is not available for annotation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RAB5
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat