- Tipo anticorpo:Primary
- Tipo di anticorpi:Sialic Acid-binding Ig-like Lectin 10
- Simbolo dell'antigene:SIGLEC10
- Clonalità:Policlonale
- Coniugazione:HRP (Horseradish Peroxidase)
- Ospite:Rabbit
- ImmunoChimica:Yes
- Isotipo:IgG
- Reattività:Human,Rat,Mouse
- Western blot:Yes
- Assorbimento incrociato:No
- Forma:liquid
- Sinomini antigene:PRO940|SLG2|SIGLEC-10
- Tampone di conservazione:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Temperatura di conservazione:Store at 4°C for 12 months
- Concentrazione:1 μg/μl
- Temperatura di spedizione:4°C
- Purificazione:Purified by Protein A
- Conf:100 µl
SIGLEC10 is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It preferentially binds to alpha2,3- or 2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. In the immune response, may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. SIGLEC10 interacts with PTPN6/SHP-1 upon phosphorylation. The protein is expressed by peripheral blood leukocytes (eosinophils, monocytes and a natural killer cell subpopulation).
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: SIGLEC10
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat