Anti-RBFOX2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-12299R-CY7
Supplier: Bioss
Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:RNA Binding Motif Protein 9
- Simbolo dell'antigene:RBFOX2
- Clonalità:Policlonale
- Coniugazione:Cy7®
- Ospite:Rabbit
- ImmunoChimica:Yes
- Isotipo:IgG
- Reattività:Human,Rat,Mouse
- Assorbimento incrociato:No
- Forma:liquid
- ID gene:23543
- Sinomini antigene:fxh|RTA|HNRBP2|Fox-2|FOX2|RBM9|HRNBP2|dJ106I20.3
- Tampone di conservazione:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Temperatura di conservazione:Store at 4°C for 12 months
- Concentrazione:1 μg/μl
- Temperatura di spedizione:4°C
- Immunogeno:100-150/390
- Purificazione:Purified by Protein A
- Conf:100 µl
Specifications
About this item
RNA-binding protein that regulates alternative splicing events by binding to 5'-UGCAUGU-3' elements. Prevents binding of U2AF2 to the 3'-splice site. Regulates alternative splicing of tissue-specific exons and of differentially spliced exons during erythropoiesis (By similarity). RNA-binding protein that seems to act as a coregulatory factor of ER-alpha.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RBFOX2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat