Order Entry
Switzerland
ContactUsLinkComponent
Anti-SPOCK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-SPOCK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Codice catalogo BOSSBS-11966R-FITC
Nome del fornitore:  Bioss
Anti-SPOCK2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Codice catalogo BOSSBS-11966R-FITC
Nome del fornitore:  Bioss

Specifiche

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Sparc/Osteonectin, Cwcv And Kazal-Like Domains Proteoglycan 2
  • Simbolo dell'antigene:
    SPOCK2
  • Clonalità:
    Policlonale
  • Coniugazione:
    FITC (Fluoresceina Isotiocionato)
  • Ospite:
    Rabbit
  • ImmunoChimica:
    Yes
  • Isotipo:
    IgG
  • Reattività:
    Human,
    Rat,
    Mouse
  • Assorbimento incrociato:
    No
  • Forma:
    liquid
  • Sinomini antigene:
    CWCV|TICN2_HUMAN.|Kazal-like domains proteoglycan 2|Sparc/osteonectin cwcv and kazal like domains proteoglycan 2|FLJ97039|SPOCK 2|Sparc/osteonectin cwcv and kazal like domains proteoglycan testican 2|TICN 2|TICN2|Testican2|KIAA0275|Testican-2|Spock2|SPARC/osteonectin
  • Tampone di conservazione:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Temperatura di conservazione:
    Store at 4°C for 12 months
  • Concentrazione:
    1 μg/μl
  • Temperatura di spedizione:
    4°C
  • Purificazione:
    Purified by Protein A
  • Conf:
    100 µl

Specifiche

Informazioni su questo articolo

The Testican family, also designated the BM-40/SPARC/osteonectin family, is composed of highly conserved, extracellular, calcium-binding, sulfate proteoglycans. Expression of Testicans is detected in a variety of tissues, but is most abundant in brain. Family members include Testican-1, Testican-2, Testican-3 and an amino-terminal splice variant of Testican-3, designated N-Tes. Most Testicans inhibit MT-MMPs, thereby inhibiting the activity of pro-MMP-2. Testican-2 is expressed in the central nervous system (CNS), with widespread expression in the olfactory bulb, cerebral cortex, thalamus, hippocampus, cerebellum and medulla, and is also found in lung and testis. Testican-2 is unique in that it actually abolishes the inhibition of MT-MMPs by other testican family members and specifically inactivates N-Tes by binding to its COOH-terminal extracellular calcium-binding domain. Testican-2 halts neurite growth from cerebellar neurons and may be involved in regu-lating the development of the CNS.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPOCK2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat