Orders LinkContactUsLinkComponent
Garbage Bins
Garbage Bins
Supplier:   LAMAPLAST
Garbage Bins
Supplier:  LAMAPLAST
Certificates
Garbage Bins
4 Options Available Below
Product Details & Documents
Bins are made of HDPE.
  • They use standard sacs 700×1000 mm
Different lid colours to separate different kind of waste.
Product Family Options
Product Information
  • Colore
    Capacità
    L×P×A
    Availability
    Price
  • blu
    50 L
    390×430×680 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 98.50
    SFr. 98.50
    1 * 1 ST
       SFr. 98.50
  • verde
    50 L
    390×430×680 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 98.50
    SFr. 98.50
    1 * 1 ST
       SFr. 98.50
  • rosso
    50 L
    390×430×680 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 98.50
    SFr. 98.50
    1 * 1 ST
       SFr. 98.50
  • giallo
    50 L
    390×430×680 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 98.50
    SFr. 98.50
    1 * 1 ST
       SFr. 98.50