Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:Brain-derived neurotrophic factor
- Simbolo dell'antigene:BDNF
- Clonalità:Monoclonale
- Clone:3C11
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Mouse
- ImmunoChimica:Yes
- ImmunoFluorescenza:Yes
- Reattività:Human,Guinea pig,Rat,Mouse
- Western blot:Yes
- Epitopo:Recombinant human mature BDNF expressed in E.coli
- Formato:Protein G purified IgG.
- Forma:Lyophilized
- ID gene:P23560 BDNF_HUMAN
- Sinomini antigene:proBDNF|Abrineurin
- Tampone di conservazione:PBS, pH 7.4, with 3% and 0.1% sodium azide as preservative.
- Temperatura di conservazione:Store lyophilized antibody at −20 °C to −80 °C protected from moisture. After reconstitution divide antibody into useful aliquots and keep aliquots at −20 °C to −80 °C for a higher stability. Working aliquots can be kept at 2-8 °C for up to 1 month.
- Temperatura di spedizione:Room (ambient) temperature.
- Immunogeno:Recombinant human mature BDNF expressed in E.coli
- Purificazione:Protein G purified.
- Taglia:100 ug
- Conf:100 µG
Specifications
About this item
BDNF belongs to the neurotrophin family and promotes the survival of neuronal populations that are all located either in the central nervous system or directly connected to it. It is a major regulator of synaptic transmission and plasticity at adult synapses in many regions of the CNS. The versatility of BDNF is emphasized by its contribution to a range of adaptive neuronal responses including long-term potentiation (LTP), long-term depression (LTD), certain forms of short-term synaptic plasticity, as well as homeostatic regulation of intrinsic neuronal excitability. The alterations in BDNF expression induced by various kinds of brain insult including stress, ischemia, seizure activity and hypoglycemia, may contribute to some pathologies such as depression, epilepsy, Alzheimer's, and Parkinson's disease. Microglia release BDNF that may contribute to neuroinflammation and neuropathic pain. SUBUNIT: Monomers and homodimers. Binds to NTRK2/TRKB. SUBCELLULAR LOCATION: Secreted protein. POst translation modification: Converted into mature BDNF by plasmin (PLG). SIMILARITY: Belongs to the NGF-beta family.
ELISA (50-100 ng/mL), WB (0.2 to 2 µg/mL), IF (2 to 20 µg/mL). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: BDNF
Clonality: Monoclonal
Clone: 3C11
Conjugation: Unconjugated
Epitope: Recombinant human mature BDNF expressed in E.coli
Host: Mouse
Isotype:
Reactivity: Human;Mouse;Rat;Guinea Pig