Baths and circulators, with KISS® controller
Informazioni su questo articolo
Heating circulation bath thermostats with either polycarbonate or stainless steel baths, which are simple to use and are suitable for a variety of applications such as: Sample temperature control, analysis, materials testing or checking food. The units offer high temperature stability and are fitted with over-temperature and low level protection suitable for Safety Class III/FL (DIN 12876) for use with flammable fluids. A powerful pressure/suction pump ensures optimum circulation and temperature homogenity in the bath and also allows temperature control of externally attached applications using a pump adapter (accessory).
- Large temperature display
- LED indicators for pump, cooling and heating
- Simple operation with menu navigation
- Auto start function for power failure
The KISS controller combines state-of-the-art technology with simple operation and stylish design. KISS models are suitable for routine tasks in research and industry and are convincing as practice oriented basic equipment. The optional PT100 sensor connection is available to display (not control) e.g. of the process temperature (only available factory fitted, at an additional charge).
Maximum pump capacities: 14 L/min, 250 mbar pressure, 10, 5 L/min, 170 mbar suction.
Certifications: Complies to safety class III/FL (DIN 12876).
Informazioni per l'ordine: Accessories included. There are also some optional accessories.
12 varianti
Product Details & Documents
Heating circulation bath thermostats with either polycarbonate or stainless steel baths, which are simple to use and are suitable for a variety of applications such as: Sample temperature control, analysis, materials testing or checking food. The units offer high temperature stability and are fitted with over-temperature and low level protection suitable for Safety Class III/FL (DIN 12876) for use with flammable fluids. A powerful pressure/suction pump ensures optimum circulation and temperature homogenity in the bath and also allows temperature control of externally attached applications using a pump adapter (accessory).
- Large temperature display
- LED indicators for pump, cooling and heating
- Simple operation with menu navigation
- Auto start function for power failure
The KISS controller combines state-of-the-art technology with simple operation and stylish design. KISS models are suitable for routine tasks in research and industry and are convincing as practice oriented basic equipment. The optional PT100 sensor connection is available to display (not control) e.g. of the process temperature (only available factory fitted, at an additional charge).
Maximum pump capacities: 14 L/min, 250 mbar pressure, 10, 5 L/min, 170 mbar suction.
Certifications: Complies to safety class III/FL (DIN 12876).
Informazioni per l'ordine: Accessories included. There are also some optional accessories.
Product Family Options
With polycarbonate bath
Informazioni sul prodotto
- TipoRange di temperaturaCost. temp.Potenza di riscaldamentoCapacitàBagno L×P×AL×P×APesoDisponibilitàPrice
- KISS 104A(15)25...100 °C±0,05 °C1,5 - 2,1 kW4 LØ 25/150 mm147×234×329 mm5 kgLog in per disponibilità1 * 1 STSFr. 2’910.00
- KISS 106A(15)25...100 °C±0,05 °C1,5 - 2,1 kW6 L130×110×150 mm147×307×330 mm5 kgLog in per disponibilità1 * 1 STSFr. 2’170.00
- KISS 108A(15)25...100 °C±0,05 °C1,5 - 2,1 kW8 L130×210×150 mm147×407×330 mm6 kgLog in per disponibilità1 * 1 STSFr. 2’210.00
- KISS 110A(15)25...100 °C±0,05 °C1,5 - 2,1 kW10 L130×310×150 mm147×507×330 mm6 kgLog in per disponibilità1 * 1 STSFr. 2’260.00
- KISS 112A(15)25...100 °C±0,05 °C1,5 - 2,1 kW15 L275×161×150 mm333×360×335 mm8 kgLog in per disponibilità1 * 1 STSFr. 2’540.00
- KISS 118A(15)25...100 °C±0,05 °C1,5 - 2,1 kW18 L275×321×150 mm333×520×335 mm8 kgLog in per disponibilità1 * 1 STSFr. 2’630.00
With stainless steel bath
Informazioni sul prodotto
- TipoRange di temperaturaCost. temp.Potenza di riscaldamentoCapacitàBagno L×P×AL×P×APesoDisponibilitàPrice
- KISS 205B(−30)25...200 °C±0,05 °C1,5 - 2,1 kW5 L195×90×150 mm178×337×355 mm9 kgLog in per disponibilità1 * 1 STSFr. 3’370.00
- KISS 208B(−30)25...200 °C±0,05 °C1,5 - 2,1 kW8,5 L230×127×150 mm290×350×375 mm10 kgLog in per disponibilità1 * 1 STSFr. 2’500.00
- KISS 212B(−30)25...200 °C±0,05 °C1,5 - 2,1 kW12 L290×152×150 mm350×375×375 mm11Log in per disponibilità1 * 1 STSFr. 2’680.00
- KISS 215B(−30)25...200 °C±0,05 °C1,5 - 2,1 kW15 L290×152×200 mm350×375×425 mm12 kgLog in per disponibilità1 * 1 STSFr. 2’800.00
- KISS 220B(−30)25...200 °C±0,05 °C1,5 - 2,1 kW20 L290×329×150 mm350×555×375 mm14 kgLog in per disponibilità1 * 1 STSFr. 2’870.00
- KISS 225B(−30)25...200 °C±0,05 °C1,5 - 2,1 kW25 L290×329×200 mm350×555×425 mm16Log in per disponibilità1 * 1 STSFr. 3’070.00