Accessories for pipette stand
Catalog # 612-5733
Supplier: BURKLE
Specifications
- Descrizione:Set for converting the stand into a pipette stand, incl. two perforated plates and one drip pan
- Conf:1
Specifications
About this item
Consisting of two perforated plates and one drip pan.