Earmuffs, helmet-mounted, Endurance, PW47
Catalog # PRTWPW47RER
Supplier: Portwest
Specifications
- Altezza ripiano:29 dB
- Colore:rosso
- Descrizione:Earmuffs, Endurance, helmet-mounted, for helmets, PS53, PW54, PW55, 29 dB, red
- Conf:1
Specifications
About this item
These helmet-mounted ear protectors made from ABS feature an excellent attenuation level.
- Can be adjusted into stand-off position when not in use
- Easy to clip on
Compatible with Portwest hemlets PS53, PW54 and PW55.
SNR: 29 dB
Certifications: EN 352-3