Autosampler vial kits, pre-assembled
Nome del fornitore: THERMO NATIONAL SCIENTIFIC
Certificati
Informazioni su questo articolo
The 8 mm vial convenience kits save time during sample preparation and reduce the risk of contamination.
Ordering information: Each kit includes 100 clear glass vials pre-assembled with 100 screw thread caps and septa.
Packaging: Packaged in convenient vial trays with clear covers or in economical polybags.
Frequently Bought Together