Replacement beakers for Goldfisch fat extractor
Codice catalogo LBCP3505300
Nome del fornitore: LABCONCO
Specifiche
- Capacità:100 ml
- Descrizione:Beakers for Goldfisch fat extractor
- Altezza:83 mm
- Conf:6
Specifiche
Informazioni su questo articolo
Beakers specifically designed for the Goldfisch Fat Extractor. Thick-walled borosilicate glass with flanged and ground tops to facilitate connection to the condenser.