Stand for mobilux® LED
N° de catalogue ESCH1511003
Fournisseur: ESCHENBACH OPTIK
Spécifications
- Descrizione:Mobase for mobilux® LED
- Environmentally Preferable:
- Conf:1
Spécifications
À propos de cet article
High quality stand for the mobilux® LED that can be used in the flat position for tremble-free reading and viewing, as well as in an upright position to free hands for carrying out work behind the lens.
- Leaves both hands free for reading and working
- Easy to use
- Maintains correct distance of use
The stand is equipped with a 1/4" thread. This enables it to be screwed on to a standard flexible tripod. The stand enables to freely adjust the distance of use and it can be used with all mobilux® LED magnifiers.