Ascorbic acid titrant soluzione
Dropping bottle, marked, 100 mL
Codice catalogo HACH2308232
Nome del fornitore: Hach
Vai a
Product Details & Documents
This titrant is used for the determination of ascorbic acid in juice based beverages, by drop-count titration.
Specifiche
Specifiche
- Tipo di conf.:Dropping bottle, marked
- Conf:100 mL