LED illumination systems, HB 100, for microscopes
Catalog # ZEIS4230119901000
Supplier: ZEISS
Specifications
- Descrizione:HBO 100 illuminator
- Conf:1
- Per:Microscopes
Specifications
About this item
HBO 100 illuminator, self-adjusting with lamp mount and collector, including heat-reflecting filter and 2 m connection cable. Automatic lamp alignment.
- Automatic lamp alignment