Soft-sided field case for portable meters, Orion™ Star series
Catalog # ORIO1210005
Supplier: Thermo Orion
Specifications
- Per:Orion Star A120, A220 + A320 series, Orion Star and Star Plus
- Descrizione:Soft-sided field case, incl. interior dividers and exterior pockets to safely organise the meter and accessories in the field
- Seq:105
- Conf:1
Specifications
About this item
This cushioned field case with interior dividers and exterior pockets protects meters, electrodes, solution and beakers under field conditions.
For use with: Orion Star A120 series portable meters; Orion Star A220 series portable meters; Orion Star A320 series portable meters; Orion 3-Star, 4-Star and 5-Star series portable meters (retired); Orion Star and Star Plus portable meters.