Hard-sided field case for portable meters, Orion™ Star A series
Catalog # ORIOSTARA-CS
Supplier: Thermo Orion
Specifications
- Per:Orion Star A120, A220 + A320 series
- Descrizione:Hard-sided field case for portable meters, holds meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeve
- Seq:105
- Conf:1
Specifications
About this item
This hard-sided field case can be used to protect, transport and store measuring equipment in the field.
The field case holds Star A series portable meter (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe and dissolved oxygen probe in storage sleeve. The electrodes and probes can remain connected to meter while in the case plus the internal separating door can be removed for cable management.
For use with: Thermo Scientific™ Orion™ Star A series portable meters (Star A320, Star A220 and Star A120 series)