Cleaning cloth, low-lint, Tork Industrial
Catalog # SCAA190578
Supplier: Essity
Specifications
- Per:W4 - Top Pak
- Confezione:80 wipes/pack, 5 packs/case
- Colore:blu
- L×L:385×385 mm
- Conf:400
Specifications
About this item
This extremely low lint cloth with anti-static properties is ideal for use with solvents when cleaning smooth surfaces. Made from 100% PP.
- Perfect for working with sensitive surfaces due to extreme the low-lint and antistatic properties
- Strong texture for excellent oil and water absorption
- Tork Easy Handling™ for easier carrying, opening and disposing of packaging
- The silicone-free cloth ensures that there are no residues on a surface
- Food contact approved
Suitable for use in clean condition areas, printing industry, precision engineering, electronics industry.
Can be used with the Tork Folded Wiper/Cloth Dispenser that protects the refills from dirt and offers one-at-a time dispensing to reduce consumption and waste.