Slide Printer, Signature
Catalog # PRMA078108
Supplier: Primera Technology Europe
Specifications
- Peso:7,62 kg
- Descrizione:Signature slide printer high pressure (HP) feature, European version with EU power cord
- L×P×A:179,8×319,5×218,4 mm
- Conf:1
Specifications
About this item
The Signature slide printer can significantly increase laborator efficiency while helping to reduce the risk of incorrect sample identification. It prints directly onto slides, eliminating handwriting errors and the need for difficult to apply, xylene-resistant labels.
- Efficient, hands-free operation
- Slides are stored in easy-to-load cartridges away from dust and other potential contaminants
- USB interface enables integration with laboratory information systems
- Chemical resistant ink ensures reliable identification of slides
With 300 dpi print resolution, the slide printed can be used to print text, graphics and logos along with high-resolution linear and 2D bar codes on every slide. The level of slides remaining in a cartridge is easily viewed through the cassette's transparent blue LED backlit case. Print speed: Up to 9 slides/minute (monochrome); 5 slides/minute (solid colours); 4 slides/minute (process colours).