Blow lamps
Blow lamp with 3 valves, Natural gas
Codice catalogo JUCHVL01621E
Nome del fornitore: JUCHHEIM Laborgerate GmbH
Vai a
Product Details & Documents
With three valves.
Height: 170 mm
Weight: 1475 g
Specifiche
- Descrizione:Blow lamp with 3 valves
- Seq:01
- Per:Natural gas