Silicone fluids
Supplier: IKA
Jump to:
About this item
3 Options Available Below
Product Details & Documents
Product Family Options
Product Information
- TipoIntervallo di temperaturaViscositàColoreConfAvailabilityPrice
- Heating fluid+20...+200 °C50 mm²/straslucido rosso10 kgLog in for availability1 * 10 kgSFr. 824.00
Accessories
Product Information
- Catalog #DescrizioneAvailabilityPrice
- IKAA20013291Laboratory reactor system LR-2.ST the High PerformerLog in for availability1 * 1 ST - SFr. 20’450.00