Total Nitrogen test kit for AQUAfast 3700 Colorimeter and AquaMate spectrophotometers
Codice catalogo ORIOACD004
Nome del fornitore: Thermo Orion
Numero CAS:
Specifiche
- Metodo chimico:Digestion
- Conf:50 Assays
- Descrizione:AQUAfast total nitrogen LR digestion tubes
- Seq:83
- Range di misura:0,5 - 25 mg/L
- Tipo:Digestion Tube
- Indice:Set of 5: 50 TN hydroxide LR digestion tubes, 50 persulfate reagent powder packs, 50 TN reagent A powder packs, 50 TN reagent B powder packs, 50 TN Acid LR/HR reaction tubes
Specifiche
Informazioni su questo articolo
Orion™ AQUAfast™ Total Nitrogen reagents in digestion tubes are designed for use with Orion™ AQUAfast™ 3700 colorimeter and Orion™ AquaMate Vis and UV-Vis spectrophotometers. Just add the sample to the digestion tube, digest sample in thermoreactor and measure.
- Reagents are fully prepared and offer convenience and added safety for user
Digestion tubes contain the precise amount of reagent, are made of special optical glass, and fit the Orion™ AQUAfast™ COD165 thermoreactor.