Water leak detectors
Nome del fornitore: EVOQUA WATER TECHNOLOGIES
Certificati
Informazioni su questo articolo
In case of leakage, a sensor activates a solenoid valve to shut down the water supply.
- Applications include, water and neutral or electrically conducting fluids
- Stainless steel internal components
- User-defined installation position
Power supply: 230 V, 50 Hz
Maximum ambient temperature: 50 °C
Delivery: Included is a controlling device, solenoid valve and 5 m connection cable.
Frequently Bought Together