Shoe covers, AlphaTec® Sure Step, model 403
Catalog # ANSEWHSSS-00403-00
Supplier: Ansell
Specifications
- Taglia scarpe:42 - 46
- Antistatico:+
- Applicazioni per abiti:General Purpose,Laboratory,Industrial
- Colore:bianco
- Seq:02
- Modello:403
- Conf:300 PAIR
- Tessuto:PP film
- Seam type:Stitched
- Peso:65 g/m²
Specifications
About this item
A disposable shoe cover that provides slip-resistance to the underside of the feet.
- Liquid resistant material
- Low linting
- Anti-static
- Elasticated opening
Weight: 65 g/m²