ESD shoe covers, MICROGARD® 2000, model 417
Catalog # ANSEWH20B-00417-02
Supplier: Ansell
Specifications
- Taglia scarpe:39 - 42
- Applicazioni per abiti:Chemical protection
- Colore:bianco
- Seq:01
- Conf:250 PAIR
Specifications
About this item
These over shoes are made from a microporous PE film with a non woven inner layer. This fabric provides low hazard liquid chemical repllence particle protection, protection from pesticides and a barrier to biological agents.
- Elasticated opening
- Bound seams
- ESD PVC sole
- Different sizes available