Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-IGLC1 Mouse Monoclonal Antibody [clone: LLC/1738]
Catalog # ABNOMAB14566
Supplier:  Abnova
Anti-IGLC1 Mouse Monoclonal Antibody [clone: LLC/1738]
Catalog # ABNOMAB14566
Supplier:  Abnova

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Native purified human Lambda Light Chain
  • Simbolo dell'antigene:
    IGLC1
  • Clonalità:
    Monoclonale
  • Clone:
    LLC/1738
  • Coniugazione:
    Non coniugato
  • Ospite:
    Mouse
  • ImmunoChimica:
    Yes
  • Isotipo:
    IgG1 kappa
  • Reattività:
    Human
  • Forma:
    Liquid
  • ID gene:
    3537
  • Tampone di conservazione:
    In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
  • Temperatura di conservazione:
    Store at 4° C.
  • Immunogeno:
    Native purified human Lambda Light Chain.
  • Purificazione:
    Protein A/G purification
  • Taglia:
    100 µg
  • Conf:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against native human Lambda Light Chain .

Type: Primary
Antigen: IGLC1
Clonality: Monoclonal
Clone: LLC/1738
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human