PEEK starter kit
Catalog # 554-1440
Supplier: VICI JOUR RESEARCH
Specifications
- Descrizione:PEEK starter kit
- Conf:1
Specifications
About this item
In LC applications involving proteins, peptides, nucleic acids, or other samples of biological origin, metal systems may interact with samples. The PEEK starter kit facilitates replacement of stainless steal tubing, fittings, ferrules, mobile phase filters and more, to create a biocompatible environment for samples and mobile phase.