Membrane kit for O₂ electrodes
Membrane kit for SE 703/706/707/706 Memosens sensors, incl. 4 × membrane body, O-ring set, and 25 ml electrolyte
N° de catalogue KNICZU0564
Fournisseur: KNICK
Aller à
Product Details & Documents
This kit includes: membrane body (4×), O-ring set (1×), and 25 ml electrolyte (1×). Suitable for SE 703/706/707/706 Memosens sensors.
Spécifications
- Descrizione:Membrane kit for SE 703/706/707/706 Memosens sensors, incl. 4 × membrane body, O-ring set, and 25 ml electrolyte