Miracloth, Calbiochem®
Catalog # 475855-1
Supplier: Merck Millipore (Calbiochem)
Specifications
- Dimensioni pori:22,00 - 25,00 µm
- Descrizione:Miracloth
- Conf:1 Roll
Specifications
About this item
Miracloth is an autoclavable, quick filtration material for gelatinous homogenates that is composed of rayon-polyester with an acrylic binder. Ideal for filtration of cleared neutralised lysates during medium- and large-scale plasmid purification to prevent contamination with loosely adherent pellet material and is useful for protoplast isolation.