Micrococcus lysodeikticus, MP Biomedicals
Micrococcus lysodeikticus , 1 g
Codice catalogo ICNA0215997201
Nome del fornitore: MP Biomedicals
Micrococcus lysodeikticus, MP Biomedicals
Codice catalogo ICNA0215997201
Nome del fornitore: MP Biomedicals
Numero CAS:
Vai a
Product Details & Documents
Lysozyme lysates harvested from cultures of Micrococcus lysodeikticus were attached to sepharose and used for affinity chromatography to isolate various bacteriolytic enzymes and it has also been used in a study to investigate the encapsulation of protein drugs in microparticles.
Specifiche
Specifiche
- Conf:1 g
- Descrizione:<i>Micrococcus lysodeikticus </i>