Rilevatore di gas, Pac® 7000
Supplier: Dräger
Certificates
About this item
Small lightweight personal monitor for industrial applications with high performance electrochemical sensors.
- Gas inflow at top and front of the monitor
- Adjustable bump test interval
- No lifetime limitation with simple battery, filter and sensor replacement
- Data logger with up to 7000 data sets - requires optional module and software for analysis
- Adjustable TWA and STEL alarms, record of peak concentration
- Automatic function test with optional Dräger Bump Test Station
Optional five year guarantee for some monitors: H₂S, O₂ and CO.
Specifications
- Catalog No:
- 118-0184
- 118-0194
- 118-0185
- 118-0195
- 118-0197
- 118-0196
- Modello:
- Pac® 7000
- Pac® 7000
- Pac® 7000
- Pac® 7000
- Pac® 7000
- Pac® 7000
- Umidità:
- 10 − 90 % RH
- 10 − 90 % RH
- 10 − 90 % RH
- 10 − 90 % RH
- 10 − 90 % RH
- 10 − 90 % RH
- Display:
- LCD
- LCD
- LCD
- LCD
- LCD
- LCD
- Registro dati:
- Up to 120 hours including concentration levels, time and date
- Up to 120 hours including concentration levels, time and date
- Up to 120 hours including concentration levels, time and date
- Up to 120 hours including concentration levels, time and date
- Up to 120 hours including concentration levels, time and date
- Up to 120 hours including concentration levels, time and date
- Allarme:
- Audible (90 dB @ 30 cm), visual (flashing LED) and vibrating
- Audible (90 dB @ 30 cm), visual (flashing LED) and vibrating
- Audible (90 dB @ 30 cm), visual (flashing LED) and vibrating
- Audible (90 dB @ 30 cm), visual (flashing LED) and vibrating
- Audible (90 dB @ 30 cm), visual (flashing LED) and vibrating
- Audible (90 dB @ 30 cm), visual (flashing LED) and vibrating
- Tipo di batteria:
- Lithium
- Lithium
- Lithium
- Lithium
- Lithium
- Lithium
- L×P×A:
- 64×25×84 mm
- 64×25×84 mm
- 64×25×84 mm
- 64×25×84 mm
- 64×25×84 mm
- 64×25×84 mm
- Peso:
- 120 g
- 120 g
- 120 g
- 120 g
- 120 g
- 120 g
- Range di temperatura:
- −30…+50 °C
- −30…+50 °C
- −30…+50 °C
- −30…+50 °C
- −30…+50 °C
- −30…+50 °C
Specifications
XXS sensor
Product Information
Frequently Bought Together