Freund's incomplete adjuvant, MP Biomedicals
Codice catalogo ICNA08642861
Nome del fornitore: MP Biomedicals
Numero CAS:
Specifiche
- Conf:50 mL
- Descrizione:Freund's incomplete adjuvant
- Environmentally Preferable:
Specifiche
Informazioni su questo articolo
Freund's incomplete adjuvant mixture contains mineral oil and Arlacel A.
- Enhances immune response in booster injections
- Water-in-oil emulsion without Mycobacterium