Loading dye solution
Codice catalogo R0611
Nome del fornitore: Thermo Fisher Scientific
Numero CAS:
Specifiche
- Conf:1 KIT
- Descrizione:Loading dye solution 6X
Specifiche
Informazioni su questo articolo
This gel loading dye contains two different dyes bromophenol blue and xylene cyanol FF for visual tracking of DNA migration during electrophoresis. The presence of glycerol ensures that the DNA in the ladder and sample forms a layer at the bottom of the well. The EDTA included in the solution binds divalent metal ions and inhibits metal-dependent nucleases.
- Two-colour tracking of DNA migration during DNA electrophoresis
- No DNA masking during gel exposure to UV light
- EDTA binds divalent metal ions and inhibits metal dependent nucleases
Used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels.