Human VEGFR2 Fc Chimera (from insect cells)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Sorgente luminosa:Insect cells
- Specie:Human
- Condizioni di conservazione:Lyophilized VEGFR-2 Fc/Chimera protein although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution FLK1 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Attività biologica:The activity of sVEGFR-2/Fc was determined by its ability to inhibit the VEGF-dependent proliferation of human umbilical vein endothelial cells. The ED50 for this effect is typically 10-30 ng/ml.
- Protein synonyms:CD309
- Protein/peptide name:VEGFR2 Fc Chimera
- Purezza:Greater than 90.0% as determined by(a)Analysis by RP-HPLC.(b)Analysis by SDS-PAGE.
- Solubilità:It is recommended to reconstitute the lyophilized VEGFR2 in sterile water not less than 100 ug/ml, which can then be further diluted to other aqueous solutions.
- Formulazione:KDR fusion protein was lyophilized from concentrated (1 mg/ml) sterile solution containing 1x PBS pH-7.4.
- Applicazioni testate:SDS-PAGE