Human angiopoietin-like 4 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Sorgente luminosa:E. coli
- Specie:Human
- Condizioni di conservazione:Store lyophilized Angiopoietin-like Protein Human recombinant at −20 °C. Aliquot the ANGPTL4 after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted ANGPTL4 °C an be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C. The lyophilized ANGPTL4 remains stable until the expiry date when stored at −20 °C.
- Protein synonyms:ANGPTL4
- Protein/peptide name:Angiopoietin-like 4
- Purezza:Greater than 95% as determined by SDS-PAGE. One-step procedure using affinity Ni-NTA chromatography.
- Solubilità:Add 0.2 uml of 0.1M Acetate buffer pH4 and let the lyophilized pellet dissolve completely. For conversion into higher pH value, we recommend intensive dilution by relevant buffer to concentration of 10ug/ml. In higher concentrations the solubility of this
- Formulazione:Sterile filtered and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH-4.
- Applicazioni testate:SDS-PAGE , HPLC , FuncS