HTNV (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Sorgente luminosa:E. coli
- Condizioni di conservazione:Two years frozen. One month in solution at room temperature.
- Protein/peptide name:HTNV
- Purezza:HTNV protein is >95% pure as determined by 10% PAGE (coomassie staining). Purified by proprietary chromatographic technique.
- Formulazione:HTNV is formulated in 1x PBS pH-7.4 and 0.5mM EDTA.
- Applicazioni testate:SDS-PAGE