HSV-2 gD
Supplier: Biorbyt
Certificates
About this item
Specifications
- Condizioni di conservazione:Five years frozen. One month in solution at room temperature.
- Protein/peptide name:HSV-2 gD
- Purezza:HSV-2 gD protein is >95% pure as determined by 10% PAGE (coomassie staining). HSV-2 gD protein was purified by proprietary chromatographic technique.
- Formulazione:25mM Tris-HCl pH 7.2, 1mM EDTA, and 50% glycerol.
- Applicazioni testate:SDS-PAGE , HPLC , FuncS