Orders LinkContactUsLinkComponent
HCV genotype-4
HCV genotype-4
Marketsource
Supplier:   Biorbyt
HCV genotype-4
Supplier:  Biorbyt
Marketsource
Certificates
2 Options Available Below
Product Details & Documents
Documents
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • BIRBORB81561-100
  • BIRBORB81561-500
  • Protein/peptide name:
  • HCV Genotype-4
  • HCV Genotype-4
  • Purezza:
  • HCV Core Genotype-4 protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV Core Genotype-4 protein was purified by proprietary chromatographic technique.
  • HCV Core Genotype-4 protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV Core Genotype-4 protein was purified by proprietary chromatographic technique.
  • Formulazione:
  • 50mM Tris-HCl, pH-8, 60mM NaCl, 10mM glutathione, 0.25% sarkosil 50% glycerol.
  • 50mM Tris-HCl, pH-8, 60mM NaCl, 10mM glutathione, 0.25% sarkosil 50% glycerol.
  • Condizioni di conservazione:
  • Five years frozen. One month in solution at room temperature.
  • Five years frozen. One month in solution at room temperature.
  • Applicazioni testate:
  • SDS-PAGE
  • SDS-PAGE
Specifications
Product Family Options
Product Information
  • Conf
    Availability
    Price
  • 100 µG
    Log in for availability
    Log in for availability
    1 * 100 µG
    1 * 100 µG
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    BIRBORB81561-100
    Protein/peptide name
    HCV Genotype-4
    Purezza
    HCV Core Genotype-4 protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV Core Genotype-4 protein was purified by proprietary chromatographic technique.
    Formulazione
    50mM Tris-HCl, pH-8, 60mM NaCl, 10mM glutathione, 0.25% sarkosil 50% glycerol.
    Condizioni di conservazione
    Five years frozen. One month in solution at room temperature.
    Applicazioni testate
    SDS-PAGE
    Marketsource
  • 500 µG
    Log in for availability
    Log in for availability
    1 * 500 µG
    1 * 500 µG
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    BIRBORB81561-500
    Protein/peptide name
    HCV Genotype-4
    Purezza
    HCV Core Genotype-4 protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV Core Genotype-4 protein was purified by proprietary chromatographic technique.
    Formulazione
    50mM Tris-HCl, pH-8, 60mM NaCl, 10mM glutathione, 0.25% sarkosil 50% glycerol.
    Condizioni di conservazione
    Five years frozen. One month in solution at room temperature.
    Applicazioni testate
    SDS-PAGE
    Marketsource