Order Entry
Switzerland
ContactUsLinkComponent
HCV-NS3-S3
HCV-NS3-S3
Catalog # BIRBORB97926-100
Supplier:  Biorbyt
CAS Number:  
HCV-NS3-S3
Catalog # BIRBORB97926-100
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Conf:
    100 µG
  • Condizioni di conservazione:
    Store at−20 °C.
  • Protein/peptide name:
    HCV-NS3-S3
  • Purezza:
    Greater than 95% by SDS-PAGE and RP-HPLC
  • Formulazione:
    Liquid. Supplied in 25 mM Tris-HCl pH 7.5, 1 mM EDTA,1.5 M urea and 50% glycerol.
  • Applicazioni testate:
    ELISA , WB

Specifications

About this item