Order Entry
Switzerland
ContactUsLinkComponent
Recombinant HIV- M
Recombinant HIV- M
Catalog # BIRBORB98890-1
Supplier:  Biorbyt
CAS Number:  
Recombinant HIV- M
Catalog # BIRBORB98890-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Conf:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Condizioni di conservazione:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Protein/peptide name:
    HIV- M
  • Purezza:
    Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
  • Formulazione:
    PBS, pH 7.4
  • Applicazioni testate:
    ELISA , WB , IHC-P , RIA

Specifications

About this item