ContactUsLinkComponent
Recombinant SCCA Ag
Catalog # BIRBORB98843-100
Supplier:  Biorbyt
CAS Number:  
Recombinant SCCA Ag
Catalog # BIRBORB98843-100
Supplier:  Biorbyt
CAS Number:  
Specifications
  • Conf:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Condizioni di conservazione:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Protein/peptide name:
    SCCA Ag
  • Purezza:
    Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
  • Formulazione:
    PBS, pH 7.4
  • Applicazioni testate:
    ELISA