Auftragserfassung
Switzerland
ContactUsLinkComponent
Human recombinant PPM1A (from E. coli)
Human recombinant PPM1A (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant PPM1A (from E. coli)
Supplier:  Biorbyt
Specifications
  • Coniugazione:
    Non coniugato
  • Protein/peptide type:
    Recombinant
  • Sorgente luminosa:
    E. coli
  • Specie:
    Human
  • Condizioni di conservazione:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    Protein Phosphatase 1A
  • UniProtKB:
    P35813 (Human)
  • Protein/peptide name:
    PPM1A
  • Purezza:
    >95% as determined by reducing SDS-PAGE.
  • Sequenza:
    MGAFLDKPKMEKHNAQGQGNGLRYGLSSMQGWRVEMEDAHTAVIGLPSGLESWSFFAVYDGHAGS QVAKYCCEHLLDHITNNQDFKGSAGAPSVENVKNGIRTGFLEIDEHMRVMSEKKHGADRSGSTAV GVLISPQHTYFINCGDSRGLLCRNRKVHFFTQDHKPSNPLEKERIQNAGGSVMIQRVNGSLAVSR ALGDFDYKCVHGKGPTEQLVSPEPEVHDIERSEEDDQFIILACDGIWDVMGNEELCDFVRSRLEV TDDLEKVCNEVVDTCLYKGSRDNMSVILICFPNAPKVSPEAVKKEAELDKYLECRVEEIIKKQGE GVPDLVHVMRTLASENIPSLPPGGELASKRNVIEAVYNRLNPYKNDDTDSTSTDDMWLEHHHHHH
  • Formulazione:
    Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4.
  • Applicazioni testate:
    SDS-PAGE
Specifications
Related Information
Frequently Bought Together