ContactUsLinkComponent
Human recombinant KPNA2 (from E. coli)
Human recombinant KPNA2 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant KPNA2 (from E. coli)
Supplier:  Biorbyt
Specifications
  • Coniugazione:
    Non coniugato
  • Protein/peptide type:
    Recombinant
  • Sorgente luminosa:
    E. coli
  • Specie:
    Human
  • Condizioni di conservazione:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    Importin Subunit Alpha2
  • UniProtKB:
    P52292 (Human)
  • Protein/peptide name:
    KPNA2
  • Purezza:
    >95% as determined by reducing SDS-PAGE.
  • Sequenza:
    MGSSHHHHHHSSGLVPRGSHMSTNENANTPAARLHRFKNKGKDSTEMRRRRIEVNVELRKAKKDD QMLKRRNVSSFPDDATSPLQENRNNQGTVNWSVDDIVKGINSSNVENQLQATQAARKLLSREKQP PIDNIIRAGLIPKFVSFLGRTDCSPIQFESAWALTNIASGTSEQTKVVVDGGAIPAFISLLASPH AHISEQAVWALGNIAGDGSVFRDLVIKYGAVDPLLALLAVPDMSSLACGYLRNLTWTLSNLCRNK NPAPPIDAVEQILPTLVRLLHHDDPEVLADTCWAISYLTDGPNERIGMVVKTGVVPQLVKLLGAS ELPIVTPALRAIGNIVTGTDEQTQVVIDAGALAVFPSLLTNPKTNIQKEATWTMSNITAGRQDQI QQVVNHGLVPFLVSVLSKADFKTQKEAVWAVTNYTSGGTVEQIVYLVHCGIIEPLMNLLTAKDTK IILVILDAISNIFQAAENLGETEKLSIMIEECGGLDKIEALQNHENESVYKASLSLIEKYFSVEE EEDQNVVPETTSEGYTFQVQDGAPGTFNF
  • Formulazione:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl,1mM DTT,20% Glycerol, pH 8.0 .
  • Applicazioni testate:
    SDS-PAGE
Specifications
Related Information
Frequently Bought Together