ContactUsLinkComponent
Human recombinant Kallikrein 8 (from HEK293 cells)
Human recombinant Kallikrein 8 (from HEK293 cells)
Nome del fornitore:  Biorbyt
Certificati
Human recombinant Kallikrein 8 (from HEK293 cells)
Nome del fornitore:  Biorbyt
Specifiche
  • Coniugazione:
    Non coniugato
  • Protein/peptide type:
    Recombinant
  • Sorgente luminosa:
    HEK293 cells
  • Specie:
    Human
  • Condizioni di conservazione:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    Kallikrein8
  • UniProtKB:
    O60259 (Human)
  • Protein/peptide name:
    Kallikrein 8
  • Purezza:
    >95% as determined by reducing SDS-PAGE.
  • Sequenza:
    QEDKVLGGHECQPHSQPWQAALFQGQQLLCGGVLVGGNWVLTAAHCKKPKYTVRLGDHSLQNKDG PEQEIPVVQSIPHPCYNSSDVEDHNHDLMLLQLRDQASLGSKVKPISLADHCTQPGQKCTVSGWG TVTSPRENFPDTLNCAEVKIFPQKKCEDAYPGQITDVMVCAGSSKGADTCQGDSGGPLVCDGALQ GITSWGSDPCGRSDKPGVYTNICRYLDWIKKIIGSKGVDHHHHHH
  • Formulazione:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, pH 8.0.
  • Applicazioni testate:
    SDS-PAGE
Specifiche
Informazioni correlate
Frequently Bought Together