Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant Kallikrein 6 (from HEK293 cells)
Human recombinant Kallikrein 6 (from HEK293 cells)
Supplier:  Biorbyt
Certificates
Human recombinant Kallikrein 6 (from HEK293 cells)
Supplier:  Biorbyt

Specifications

  • Coniugazione:
    Non coniugato
  • Protein/peptide type:
    Recombinant
  • Sorgente luminosa:
    HEK293 cells
  • Specie:
    Human
  • Condizioni di conservazione:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Protein synonyms:
    Kallikrein6
  • UniProtKB:
    Q92876 (Human)
  • Protein/peptide name:
    Kallikrein 6
  • Purezza:
    >95% as determined by reducing SDS-PAGE.
  • Sequenza:
    EEQNKLVHGGPCDKTSHPYQAALYTSGHLLCGGVLIHPLWVLTAAHCKKPNLQVFLGKHNLRQRE SSQEQSSVVRAVIHPDYDAASHDQDIMLLRLARPAKLSELIQPLPLERDCSANTTSCHILGWGKT ADGDFPDTIQCAYIHLVSREECEHAYPGQITQNMLCAGDEKYGKDSCQGDSGGPLVCGDHLRGLV SWGNIPCGSKEKPGVYTNVCRYTNWIQKTIQAKVDHHHHHH
  • Formulazione:
    Supplied as a 0.2 um filtered solution of 5mM HCl, 150mM NaCl.
  • Applicazioni testate:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together