ContactUsLinkComponent
Human Recombinant CCL3L1 (from HEK293 cells)
Human Recombinant CCL3L1 (from HEK293 cells)
Supplier:  Biorbyt
Certificates
Human Recombinant CCL3L1 (from HEK293 cells)
Supplier:  Biorbyt
Specifications
  • Coniugazione:
    Non coniugato
  • Protein/peptide type:
    Recombinant
  • Sorgente luminosa:
    HEK293 cells
  • Specie:
    Human
  • Condizioni di conservazione:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    CC Motif Chemokine 3Like 1
  • UniProtKB:
    P16619 (Human)
  • Protein/peptide name:
    CCL3L1
  • Purezza:
    >95% as determined by reducing SDS-PAGE.
  • Sequenza:
    APLAADTPTACCFSYTSRQIPQNFIADYFETSSQCSKPSVIFLTKRGRQVCADPSEEWVQKYVSD LEPSAVDHHHHHH
  • Formulazione:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2.
  • Applicazioni testate:
    SDS-PAGE
Specifications
Related Information
Frequently Bought Together