Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant FCRL1 (from HEK293 cells)
Human recombinant FCRL1 (from HEK293 cells)
Nome del fornitore:  Biorbyt
Certificati
Human recombinant FCRL1 (from HEK293 cells)
Nome del fornitore:  Biorbyt

Specifiche

  • Coniugazione:
    Non coniugato
  • Protein/peptide type:
    Recombinant
  • Sorgente luminosa:
    HEK293 cells
  • Specie:
    Human
  • Condizioni di conservazione:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Fc ReceptorLike 1
  • UniProtKB:
    Q96LA6 (Human)
  • Protein/peptide name:
    FCRL1
  • Purezza:
    >95% as determined by reducing SDS-PAGE.
  • Sequenza:
    AELFLIASPSHPTEGSPVTLTCKMPFLQSSDAQFQFCFFRDTRALGPGWSSSPKLQIAAMWKEDT GSYWCEAQTMASKVLRSRRSQINVHRVPVADVSLETQPPGGQVMEGDRLVLICSVAMGTGDITFL WYKGAVGLNLQSKTQRSLTAEYEIPSVRESDAEQYYCVAENGYGPSPSGLVSITVRIPVSRPILM LRAPRAQAAVEDVLELHCEALRGSPPILYWFYHEDITLGSRSAPSGGGASFNLSLTEEHSGNYSC EANNGLGAQRSEAVTLNFTVPTGARSNVDHHHHHH
  • Formulazione:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 1mM EDTA, pH 7.4.
  • Applicazioni testate:
    SDS-PAGE

Specifiche

Informazioni correlate

Frequently Bought Together